Thermo Scientific™
Shandon™ Kwik-Diff™ Stains
Related applications:
Anatomical Pathology
Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Thermo Scientific™ Shandon™ Kwik-Diff™ Stains
Back to top
  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori
Includes:
Reagent 1: Fixative; Reagent 2: Eosin; Reagent 3: Methylene Blue