Thermo Scientific™
Shandon™ Wright-Giemsa Stain Kit
Related applications:
Anatomical Pathology
Stain using eosinates of polychrome methylene blue, which yield rich color, with the Thermo Scientific™ Shandon™ Wright-Giemsa Stain Kit.
Back to top
  • Produces an intense differential stain for blood smears, bone marrows, mast cells and FNA's
  • Demonstrates micro-organisms such as Microfilaria, Babesia, Haemobartonella, Giardia, Pneumocystis and Helicobacter