Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™
Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™
Fisher Chemical™

Ammonia Solution, 35%, Extra Pure, SLR, d=0.88, Fisher Chemical™

H5NO, CAS Number-1336-21-6, 2.5L, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Catalog number A-3240-PB15
also known as A/3240/PB15
Price (USD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
CAS1336-21-6
ColorColorless
Formula Weight35.05g/mol
MDL Number11418
Molecular FormulaH5NO
View more
H5NO, CAS Number-1336-21-6, 2.5L, Colorless, 35.05g/mol, 11418, Plastic Coated Glass bottle, 12, Liquid