Detection Method: | Colorimetric |
---|---|
Label or Dye: | Digoxigenin, HRP (Horseradish Peroxidase) |
Substrate: | AEC |
Shipping Condition: | Wet Ice |
Substrate Type: | HRP (Horseradish Peroxidase) Substrate |
Target Molecule: | DNA |
Target Specificity: | Not Target-Specific |
Product Line: | REMBRANDT® |