2,6-Dichloroindophenol sodium salt hydrate
2,6-Dichloroindophenol sodium salt hydrate
Thermo Scientific Chemicals

2,6-Dichloroindophenol sodium salt hydrate

CAS: 1266615-56-8 | C12H6Cl2NNaO2 | 290.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10107.14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS1266615-56-8
IUPAC Namesodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Molecular FormulaC12H6Cl2NNaO2
InChI KeyCVSUAFOWIXUYQA-UHFFFAOYSA-M
SMILES[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to black
CommentSuitable for ascorbic acid determination
Specific Absorbance (E 1%/1cm)≥700 @ lambda max
lambda-max (UV-Vis)638-645nm (50% ethanol + 2ml 1M sodium carbonate soln)
FormPowder
View more
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dichloroindophenol sodium salt hydrate is suitable for vitamin C determination. It acts as a redox dye to investigate the efficiency of gold nanoparticles (Au-NP) for the enzymatic activity of glucose oxidase. It is also used to compose UVB specific dosimeter and to measure the rate of photosynthesis. It serves as a good reducing agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Redox indicator: Anal. Biochem., 101, 421 (1980).
  2. Chiua, S-H.; Urban, P .L. Fusion of microlitre water-in-oil droplets for simple, fast and green chemical assays. Analyst 2015, 40 (15), 5145-5151.
  3. Phillips, T. W.; Bannock, J. H.; deMello,J. C. Microscale extraction and phase separation using a porous capillary. Lab Chip. 2015, 15 (14), 2960-2967.