2,3-Dichlorobenzaldehyde, 98%
2,3-Dichlorobenzaldehyde, 98%
2,3-Dichlorobenzaldehyde, 98%
Thermo Scientific Chemicals

2,3-Dichlorobenzaldehyde, 98%

CAS: 6334-18-5 | C7H4Cl2O | 175.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A10118.18
also known as A10118-18
Price (TWD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS6334-18-5
IUPAC Name2,3-dichlorobenzaldehyde
Molecular FormulaC7H4Cl2O
InChI KeyLLMLNAVBOAMOEE-UHFFFAOYSA-N
SMILESClC1=CC=CC(C=O)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder or fused/lumpy solid
Free acid (titration)≤1.5%
Melting Point (clear melt)59.5-65.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only