4-(Bromomethyl)benzenesulfonyl chloride, 95%
4-(Bromomethyl)benzenesulfonyl chloride, 95%
4-(Bromomethyl)benzenesulfonyl chloride, 95%
Thermo Scientific Chemicals

4-(Bromomethyl)benzenesulfonyl chloride, 95%

CAS: 66176-39-4 | C7H6BrClO2S | 269.537 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A10120.14
also known as A10120-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS66176-39-4
IUPAC Name4-(bromomethyl)benzene-1-sulfonyl chloride
Molecular FormulaC7H6BrClO2S
InChI KeyQXTQWYZHHMQSQH-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥94.0 to ≤106.0%
Identification (FTIR)Conforms
Melting Point (clear melt)67.0-80.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only