1,4-Ditioeritritol, 99 %, Thermo Scientific Chemicals
1,4-Ditioeritritol, 99 %, Thermo Scientific Chemicals
1,4-Ditioeritritol, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,4-Ditioeritritol, 99 %, Thermo Scientific Chemicals

CAS: 6892-68-8 | C4H10O2S2 | 154.24 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Número de catálogo A10138.14
también denominado A10138-14
Precio (USD)
-
Cantidad:
25 g
Pedido a granel o personalizado
Identificadores químicos
CAS6892-68-8
IUPAC Name(2R,3S)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-ZXZARUISSA-N
SMILESO[C@@H](CS)[C@H](O)CS
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White
Assay (Silylated GC)≥98.5%
Melting Point (clear melt)80.0-86.0?C
FormCrystals or powder or crystalline powder
Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
Se utiliza como reactivo para mantener grupos –SH en estado reducido y reduce cuantitativamente los disulfuros.

Solubilidad
Soluble en agua (15,4 mg/ml) y metanol.

Notas
Sensible al aire. Almacenar alejado del aire. Mantener en un recipiente bien sellado. Almacenar alejado de agentes reductores. Almacenar en gas inerte seco.
RUO – Research Use Only

General References:

  1. Morgane Riou; David Stroebel; J Michael Edwardson; Pierre Paoletti. An alternating GluN1-2-1-2 subunit arrangement in mature NMDA receptors. Plosone. 2012, 7 (4), 35134 .
  2. Yang-Rae Kim et. al. Highly sensitive gold nanoparticle-based colorimetric sensing of mercury(II) through simple ligand exchange reaction in aqueous media. ACS Applied Materials & Interfaces. 2010, 2 (1), 292-295.
  3. Protective agent for preventing oxidation of thiol groups and reagent for reduction of disulfide groups in proteins: Biochemistry, 3, 480 (1964); 11, 2939 (1972); J. Biol. Chem., 243, 716 (1968); compare 1,4-Dithio-DL-threitol, A15797.