Thermo Scientific Chemicals

Calcium fluoride, 97%, Thermo Scientific Chemicals

Catalog number: A10176.0I
5000 g, Each
Thermo Scientific Chemicals

Calcium fluoride, 97%, Thermo Scientific Chemicals

Catalog number: A10176.0I
5000 g, Each
Quantity
Catalog number: A10176.0I
also known as A10176-0I
Price (USD)
Price: 336.00
Online price: 285.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7789-75-5
IUPAC Name
calcium difluoride
Molecular Formula
CaF2
InChI Key
WUKWITHWXAAZEY-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ca++]
Appearance (Color)
White to grey
Form
Powder
Loss on Drying
<2.5%
Assay from Supplier's CofA
≥96.0%

Description

Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium fluoride is used to manufacture optical components such as windows and lenses, used in thermal imaging systems, spectroscopy, and excimer lasers. In combination with KF, it is a useful fluorinating agent for conversion of other alkyl halides to fluorides. It may be used in the fabrication of microfluidic capillary electrophoresis (CE) devices.

Solubility
Soluble in ammonium salts. Soluble in water (slightly).

Notes
Store at room temperature. Separated from mineral acids. Protect from humidity and water. Incompatible with moisture and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text