n-Propyl 4-hydroxybenzoate, 99+%
n-Propyl 4-hydroxybenzoate, 99+%
n-Propyl 4-hydroxybenzoate, 99+%
Thermo Scientific Chemicals

n-Propyl 4-hydroxybenzoate, 99+%

CAS: 94-13-3 | C10H12O3 | 180.203 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1000 g
5000 g
產品號碼 A10180.0B
亦稱為 A10180-0B
價格 (TWD)
-
Quantity:
1000 g
Request bulk or custom format
化學識別
CAS94-13-3
IUPAC Namepropyl 4-hydroxybenzoate
Molecular FormulaC10H12O3
InChI KeyQELSKZZBTMNZEB-UHFFFAOYSA-N
SMILESCCCOC(=O)C1=CC=C(O)C=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to pale cream
Melting Point (clear melt)95-100?C
FormPowder or crystalline powder
Identification (FTIR)Conforms
Assay (GC)≥99.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
n-Propyl 4-hydroxybenzoate is used as antimicrobial preservatives in pharmaceuticals, foods, cosmetics and shampoos. It is widely used as preservatives in personal care products like creams, lotions and bath products. Further, it is used as a food additive. Its derivative, sodium propyl p-hydroxybenzoate is used as an anti-fungal preservation agent.

Solubility
Soluble in propylene glycol, ethanol, ether, acetone, methanol anddimethyl sulfoxide. Slightly soluble in glycerin. Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali metals.
RUO – Research Use Only

General References:

  1. Barberis, C. L.; Landa, M. F.; Barberis, M. G.; Giaj-Merlera, G.; Dalcero, A. M.; Magnoli, C. E. Hydrolytic enzymes production by Aspergillus section Nigri in presence of butylated hydroxyanisole and propyl paraben on peanut meal extract agar. Rev. Iberoam. Micol. 2014, 31 (2), 131-136.
  2. Albunia, A. R.; Guerra, G. Spectroscopic Investigation of Guest-Guest Interactions in the Nanoporous-Crystalline delta and epsilon Forms of Syndiotactic Polystyrene. J. Phys. Chem. C 2014, 118 (22), 11774-11783.