3,5-Bis(trifluoromethyl)phenol, 97%
3,5-Bis(trifluoromethyl)phenol, 97%
3,5-Bis(trifluoromethyl)phenol, 97%
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenol, 97%

CAS: 349-58-6 | C8H4F6O | 230.109 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10183.14
also known as A10183-14
Price (EUR)
308,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
308,00
Each
Chemical Identifiers
CAS349-58-6
IUPAC Name3,5-bis(trifluoromethyl)phenol
Molecular FormulaC8H4F6O
InChI KeyODSXJQYJADZFJX-UHFFFAOYSA-N
SMILESOC1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0% (U.S. specification)
Assay (GC)≥96.0%
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only