Tetraethylammonium tetrafluoroborate, 99%
Tetraethylammonium tetrafluoroborate, 99%
Tetraethylammonium tetrafluoroborate, 99%
Thermo Scientific Chemicals

Tetraethylammonium tetrafluoroborate, 99%

CAS: 429-06-1 | C8H20BF4N | 217.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
100 g
250 g
Catalog number A10211.06
also known as A10211-06
Price (USD)
33.60
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
33.60
Each
Chemical Identifiers
CAS429-06-1
IUPAC Nametetraethylazanium; tetrafluoroboranuide
Molecular FormulaC8H20BF4N
InChI KeyXJRAKUDXACGCHA-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofABoron: ≥ 4.9% (U.S. specification)
Water Content (Karl Fischer Titration)≤0.8% (non-U.S. specification)
Assay (unspecified)≥98.5% by C & N EA (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Appearance (Color)White
View more
Tetraethylammonium tetrafluoroborate is used as a phase transfer catalyst in organic synthesis. It finds application as an electrolyte additive in the capillary electrophoretic separation of polyphenols found in grape seed extracts. It is also used to prepare other tetrabutylammonium salts in aqueous solutions. It is involved in the synthesis of conducting poly(thiophenes).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium tetrafluoroborate is used as a phase transfer catalyst in organic synthesis. It finds application as an electrolyte additive in the capillary electrophoretic separation of polyphenols found in grape seed extracts. It is also used to prepare other tetrabutylammonium salts in aqueous solutions. It is involved in the synthesis of conducting poly(thiophenes).

Solubility
Soluble in water, alcohol and acetonitrile.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Electrolyte for electro-organic synthesis, e.g. in the o- and p- hydroxylation of simple aromatic ketones, acids and esters by anodic oxidation in TFA: J. Chem. Soc., Chem. Commun., 262 (1975). In CH3CN, the products are the corresponding o- and p-acetamido compounds (cf Ritter reaction).
  2. Use as a supporting electrolyte and F- source for electrochemical fluorination has also been reported: J. Electroanal. Chem., 43, 318 (1973).
  3. Sivaraman, P.; Shashidhara, K.; Thakur, A. P.; Samui, A. B.; Bhattacharyya, A. R. Nanocomposite solid polymer electrolytes based on polyethylene oxide, modified nanoclay, and tetraethylammonium tetrafluoroborate for application in solid-state supercapacitor. Polym. Eng. Sci. 2015, 55 (7), 1536-1545.
  4. Schütter, C.; Ramirez-Castro, C.; Oljaca, M.; Passerini, S.; Winter, M.; Balducci, A. Activated Carbon, Carbon Blacks and Graphene Based Nanoplatelets as Active Materials for Electrochemical Double Layer Capacitors: A Comparative Study. J. Electrochem. Soc. 2015, 162 (1), A44-A51.