Imidazole, 99%
Imidazole, 99%
Imidazole, 99%
Thermo Scientific Chemicals

Imidazole, 99%

CAS: 288-32-4 | C3H4N2 | 68.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A10221.36
Price (EUR)
91,60
Each
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (EUR)
91,60
Each
Add to cart
Chemical Identifiers
CAS288-32-4
IUPAC Name1H-imidazole
Molecular FormulaC3H4N2
InChI KeyRAXXELZNTBOGNW-UHFFFAOYSA-N
SMILESN1C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
CommentPurchased in the U.K. and in other countries
Water Content (Karl Fischer Titration)≤0.5%
Assay from Suppliers CofA≥98.5 to ≤101.5% (Acidimetric) (non-U.K. specification)
Appearance (Color)White to pale cream to pale yellow
FormCrystals or flakes or powder or crystalline flakes or crystalline powder
View more
Imidazole is used as a buffer in the range of pH 6.2-7.8. It is also an histamine antagonist. It acts as a chelator and forms complexes with various divalent cations. It is used as a corrosion inhibitor on certain transition metals such as copper. Its derivatives, like polybenzimidazole (PBI), act as fire retardants. It finds application in photography and electronics. Imidazole salts are used as ionic liquids and precursors to stable carbenes. Imidazole derivatives like ketoconazole, miconazole and clotrimazole are involved in the treatment of various systemic fungal infections. It is a part of the theophylline molecule, present in tea leaves and coffee beans, which stimulates the central nervous system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole is used as a buffer in the range of pH 6.2-7.8. It is also an histamine antagonist. It acts as a chelator and forms complexes with various divalent cations. It is used as a corrosion inhibitor on certain transition metals such as copper. Its derivatives, like polybenzimidazole (PBI), act as fire retardants. It finds application in photography and electronics. Imidazole salts are used as ionic liquids and precursors to stable carbenes. Imidazole derivatives like ketoconazole, miconazole and clotrimazole are involved in the treatment of various systemic fungal infections. It is a part of the theophylline molecule, present in tea leaves and coffee beans, which stimulates the central nervous system.

Solubility
Miscible with water, ethanol, ether, acetone, chloroform, pyridine and methanol. Slightly miscible with benzene and petroleum ether.

Notes
Incompatible with acids, acid anhydrides and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Nucleophilic catalyst for many acylation and silylation reactions, compare 4-(Dimethyl amino) pyridine, A13016.
  2. For use in: silylation of 1,3-diketones, see Hexamethyl disilazane, A15139; introduction of the TBDMS and TBDPS groups; see: tert-Butyl dimethyl chlorosilane, A13064, and tert-Butyl diphenyl chlorosilane, A12721, respectively.
  3. In combination with triphenylphosphine and iodine, vic-diols are converted to alkenes: Synthesis, 469 (1979), and alcohols to alkyl iodides: Synth. Commun., 20, 1473 (1990).
  4. With 2 moles of an aroyl halide gives, after hydrolysis, good yields of 2-aroylimidazoles: Synthesis, 675 (1978).
  5. Sankar, M.; Ajithkumar, T. G.; Sankar, G.; Manikandan, P. Supported imidazole as heterogeneous catalyst for the synthesis of cyclic carbonates from epoxides and CO 2. Catal. Commun. 2015, 59, 201-205.
  6. Hart, K. R.; Sottos, N. R.; White, S. R. Repeatable self-healing of an epoxy matrix using imidazole initiated polymerization. Polymer 2015, 67, 174-184.