Thermo Scientific Chemicals

trans-Stilbene, 98%, Thermo Scientific Chemicals

Catalog number: A10232.36
500 g, Each
Thermo Scientific Chemicals

trans-Stilbene, 98%, Thermo Scientific Chemicals

Catalog number: A10232.36
500 g, Each
Quantity
Catalog number: A10232.36
Price (USD)
Price: 645.00
Online price: 548.65
Your price:
Quantity
-

Chemical Identifiers

CAS
103-30-0
IUPAC Name
[(1E)-2-phenylethenyl]benzene
Molecular Formula
C14H12
InChI Key
PJANXHGTPQOBST-VAWYXSNFSA-N
SMILES
C(=C/C1=CC=CC=C1)\C1=CC=CC=C1
Identification (FTIR)
Conforms
Appearance (Color)
White to cream
Form
Crystals or powder or crystalline powder or fused solid
Assay (GC)
≥97.5%
Melting Point (clear melt)
121.0-127.0?C

Description

trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-Stilbene is used in the manufacturing of dye lasers, optical brighteners, non-steroidal synthetic estrogens such as diethylstilbestrol, fosfestrol and dienestrol. It is used as a phosphor and a scintillator. It is also involved in transition metal catalyzed asymmetric epoxidation and dihydroxylation. Further, it is used as raw material in organic synthesis.

Solubility
Soluble in benzene and ether. Insoluble in water.

Notes
Air and light sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text