Gibco™ Applications liées: Antibody & Cell Culture Production | Mammalian Cell Culture
Error lors du chargement de votre contenu!
| Numéro de catalogue |
Choisir votre mod�le de prix
Taille unitaire
|
Prix ({{currency}}) | Disponibilité | Qté | |
|---|---|---|---|---|---|
| {{product.sku}} {{product.sku}} également connu comme {{product.formattedSku}} |
|
— |
| Cell Line: | CHO |
|---|---|
| Classification: | Animal Origin-Free, Chemically Defined, Protein-Free, Serum-Free |
| Comments: | Chemically Defined Nutrient Feeding Supplement |
| Culture Type: | Suspension Cell Culture |
| Endotoxin Level: | Low |
| Form: | Liquid |
| Format: | Bottle(s) |
| Glutamine: | No Glutamine |
| Included Antibiotics: | No antibiotics |
| Other Additives: | No Insulin |
| Phenol Red Indicator: | No Phenol Red |
| Product Line: | Gibco® |
| Product Size: | 1,000 mL |
| Purity or Quality Grade: | Cell Culture Grade, Qualified for Further Manufacturing, Research Grade |
| Serum Level: | Serum-Free |
| Sodium Bicarbonate Buffer: | Sodium Bicarbonate |
| Sodium Pyruvate Additive: | Sodium Pyruvate |
| Sterility: | Sterile |
| Tests Performed: | pH |
| Volume (Metric): | 1 L |
| Green Features: | Sustainable packaging |
| Shipping Condition: | Room Temperature |
| Numéro de catalogue: | NP0001 |
| Taille | 500 mL |
| Prix | EUR 79.50 |
| Ajouter aux favoris | |
| Numéro de catalogue: | NP0004 |
| Taille | 250 µL |
| Prix | EUR 14.52 |
| Ajouter aux favoris | |
| Numéro de catalogue: | LC2672 |
| Taille | 500 mL |
| Prix | EUR 40.77 |
| Ajouter aux favoris | |