Thermo Scientific Chemicals

Cytidine, 99%

Catalog number: A10261.18
50 g, Each
Thermo Scientific Chemicals

Cytidine, 99%

Catalog number: A10261.18
50 g, Each
Quantity
Catalog number: A10261.18
also known as A10261-18
Price (USD)
Quantity
-

Chemical Identifiers

CAS
65-46-3
IUPAC Name
4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular Formula
C9H13N3O5
InChI Key
UHDGCWIWMRVCDJ-UHFFFAOYNA-N
SMILES
NC1=NC(=O)N(C=C1)C1OC(CO)C(O)C1O
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5% (non-U.S. specification)
Assay (HPLC)
≥98.5% (non-U.S. specification)
Assay from Suppliers CofA
≥98.5% (HPLC: U.S. specification)

Description

Cytidine is used to synthesize enzyme inhibitors, antiviral agents, and anticancer agents. And also used as constituent of nucleic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text