2-Hydroxy-5-nitrobenzaldehyde, 98%
2-Hydroxy-5-nitrobenzaldehyde, 98%
2-Hydroxy-5-nitrobenzaldehyde, 98%
Thermo Scientific Chemicals

2-Hydroxy-5-nitrobenzaldehyde, 98%

CAS: 97-51-8 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10264.14
also known as A10264-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS97-51-8
IUPAC Name2-hydroxy-5-nitrobenzaldehyde
Molecular FormulaC7H5NO4
InChI KeyIHFRMUGEILMHNU-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1C=O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow to orange
Assay (GC)≥97.5%
FormPowder
Melting Point (clear melt)122.0-129.0?C
2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxy-5-nitrobenzaldehyde is used in the synthetic preparation of coumarin (C755380), an pharmaceutic aid that is found in tonka beans, lavender oil, woodruff, sweet clover.

Solubility
Soluble in water (partly).

Notes
Air & Light Sensitive,. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. W J Arion et. al. Chlorogenic acid and hydroxynitrobenzaldehyde: new inhibitors of hepatic glucose 6-phosphatase. Archives of Biochemistry and Biophysics. 1997, 339,(2), 315-322.
  2. H. Tanak; M. Macit; M. Yavuz; and S. Isik. 2-Hydroxy-5-nitrobenzaldehyde. Acta Crystallogr. Sect. E: Struct. Rep. Online. 2009, 65,(12),