Thermo Scientific Chemicals

Pinacolone, 97%, Thermo Scientific Chemicals

Catalog number: A10289.36
500 g, Each
Thermo Scientific Chemicals

Pinacolone, 97%, Thermo Scientific Chemicals

Catalog number: A10289.36
500 g, Each
Quantity

Chemical Identifiers

CAS
75-97-8
IUPAC Name
3,3-dimethylbutan-2-one
Molecular Formula
C6H12O
InChI Key
PJGSXYOJTGTZAV-UHFFFAOYSA-N
SMILES
CC(=O)C(C)(C)C
Appearance (Color)
Clear, colourless to yellow
Form
liquid
Refractive Index
1.3945 - 1.3995 @20?C
Assay (GC)
> 96.0%

Description

Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pinacolone is a precursor to triazolylpinacolone, triadimefon, paclobutrazol, uniconazole and metribuzin. Furthermore, it acts as an intermediate for biologically active products such as antibacterial, antifungal, antiviral and antituberculous products. It is an important ketone in organic chemistry and participates in condensation, hydrogenation and reductive amination reactions.

Solubility
Miscible with water, chloroform, ethanol, ether, acetone and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text