SuperScript® hESC Full-Length Normalized cDNA Library, BG01 - FAQs
SuperScript® hESC Full-Length Normalized cDNA Library, BG01 - FAQs
View additional product information for SuperScript® hESC Full-Length Normalized cDNA Library, BG01 - FAQs (A1030301)
1 product FAQs found
What is the best way to prepare a plasmid prep of an Invitrogen premade library in order to get the highest representation of clones for transfection of mammalian cells?
The key to minimizing a change in library representation is to put the library through as few cell divisions as possible. Initiate a culture with as much of the frozen library as possible. Growing up the library from a very small number of starting cells is not ideal since a large number of cell divisions will be required to reach the appropriate density. Do not amplify the library because this will also change the representation.