L-Methionine, 98+%
L-Methionine, 98+%
L-Methionine, 98+%
Thermo Scientific Chemicals

L-Methionine, 98+%

L-Methionine (CAS # 63-68-3) is an essential amino acid that contains sulphur. L-Methionine is involved in cellular detoxification in several species. | CAS: 63-68-3 | C5H11NO2S | 149.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
2500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 A10318.36
亦稱為 A10318-36
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS63-68-3
IUPAC Name(2S)-2-amino-4-(methylsulfanyl)butanoic acid
Molecular FormulaC5H11NO2S
InChI KeyFFEARJCKVFRZRR-BYPYZUCNSA-N
SMILESCSCC[C@H](N)C(O)=O
檢視更多
規格Specification Sheet規格表
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Appearance (Color)White
CommentMaterial sourced in the U.S. and in other countries
Identification (FTIR)Conforms (non-U.S. sourced material)
檢視更多
L-Methionine iserves as a chelating agent for heavy metals, as a flavoring agent and nutritional supplement for foods. It acts as a feed additive, vegetable oil enrichment and as a single cell protein. In addition to this, it is used as a hepatoprotectant and also acts as a lipotropic agent and prevents excess fat buildup in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Methionine is an essential amino acid required for tissue repair and maintenance

• L-Methionine affects absorption of selenium and zinc

• L-Methionine can assist cellular detoxification by chelating lead and mercury

• L-Methionine exhibitis properties associated with anti-aging and can also prevent the buildup of fat in the liver

Application

• L-Methionine is appropriate for use in cell culture and biochemistry laboratory applications

• It is useful for the laboratory study of methionine pathways in the cell relating to aging, degeneration, and metabolism

RUO – Research Use Only

General References:

  1. Bárcena C, López-Otín C, Kroemer G. Methionine restriction for improving progeria: another autophagy-inducing anti-aging strategy? Autophagy. 2019 Mar;15(3):558-559. doi: 10.1080/15548627.2018.1533059. Epub 2018 Oct 18. PMID: 30304972; PMCID: PMC6351129.
  2. Mota-Martorell N, Jové M, Berdún R, Pamplona R. Plasma methionine metabolic profile is associated with longevity in mammals. Commun Biol. 2021 Jun 11;4(1):725. doi: 10.1038/s42003-021-02254-3. PMID: 34117367; PMCID: PMC8196171.