Thermo Scientific™

Molecular sieves, 13X, powder, Thermo Scientific™

250g, Each
Thermo Scientific™

Molecular sieves, 13X, powder, Thermo Scientific™

250g, Each
Quantity
Catalog number: A10378.30
also known as A10378-30
Price (USD)
Price: 30.70 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
63231-69-6
Appearance (Color)
White to brown
Moisture holding capacity
≥25.0%
Form
Powder
Capacity
(Carbon dioxide, CO₂, adsorption): ≥18.0%
Comment
Nominal pore size: 10 Angstroms

Description

They are used as catalyze carrier, co adsorption of CO2 and H2O and H2O, H2O and H2S, as desiccant for medical and air compressor systems. They are normally used in gas phase and liquid phase applications. It finds its a major applications in commercial gas drying, air plant feed purification (simultaneous H2O and CO2 removal) and liquid hydrocarbon/natural gas sweetening (H2S and mercaptan removal).

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

RUO – Research Use Only

Figures