Thermo Scientific Chemicals

Pyridine N-oxide, 95%, Thermo Scientific Chemicals

Catalog number: A10419.36
500 g, Each
Thermo Scientific Chemicals

Pyridine N-oxide, 95%, Thermo Scientific Chemicals

Catalog number: A10419.36
500 g, Each
Quantity
Catalog number: A10419.36
also known as A10419-36
Price (USD)
Price: 252.00
Online price: 217.65
Your price:
Quantity
-

Chemical Identifiers

CAS
694-59-7
IUPAC Name
pyridin-1-ium-1-olate
Molecular Formula
C5H5NO
InChI Key
ILVXOBCQQYKLDS-UHFFFAOYSA-N
SMILES
[O-][N+]1=CC=CC=C1
Form
Crystals or powder or crystalline powder or lumps or chunks or fused solid
Melting Point (clear melt)
57.0-69.0?C
Assay (GC)
≥94.0%
Water Content (Karl Fischer Titration)
≤2.5%
Appearance (Color)
White to cream to yellow to pale brown

Description

Pyridine N-oxide is used in the treatment of Kayser's disease. It is widely used in chemical industry. It can act as catalytic agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Storage under a nitrogen blanket has been recommended. Store under 2-8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text