Thermo Scientific Chemicals

L-Menthol, 99%, Thermo Scientific Chemicals

Catalog number: A10474.0B
1000 g, Each
Thermo Scientific Chemicals

L-Menthol, 99%, Thermo Scientific Chemicals

Catalog number: A10474.0B
1000 g, Each
Quantity
Catalog number: A10474.0B
also known as A10474-0B
Price (USD)
Price: 293.00
Online price: 249.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2216-51-5
IUPAC Name
(1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol
Molecular Formula
C10H20O
InChI Key
NOOLISFMXDJSKH-KXUCPTDWSA-N
SMILES
CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
Assay (GC)
≥98.5%
Appearance (Color)
Colorless
Melting Point (clear melt)
39.0-46.0?C
Optical Rotation
-49.7 ± 0.5? (C=10 in ethanol)
Form
Crystals or powder or crystalline powder

Description

L-Menthol is used as a cooling agent that strongly activates TRPM8.(Transient Receptor Potential Cation Channel, Subfamily M, Member 8 is a Protein Coding gene). It is used as analgesic (topical), antipruritic agent. It is used as: refreshing agent, food flavor, cool and antipruritic drug, carminative drug. Menthol crystals is used for pers onal care and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 100% ethanol, methanol (100 mg/ml), ether, and chloroform. Insoluble in water.

Notes
Store at room temperature. Incompatible with Strong oxidizing agents, phenols, potassium permanganate, chromium trioxide, chloral hydrate, thymol, pyrogallol, resorcinol, camphor, butylchloral hydrate, betanaphthol, exalgine.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text