APEX™ Alexa Fluor™ 594 Antibody Labeling Kit - FAQs
APEX™ Alexa Fluor™ 594 Antibody Labeling Kit - FAQs
View additional product information for APEX™ Antibody Labeling Kits - FAQs (A10474)
1 product FAQs found
I need to label my tissue section with two mouse primary antibodies. Do I have to make direct conjugates of the antibodies to use them?
Check the isotype of the two antibodies to see if they are different. If so, we offer isotype-specific goat anti-mouse secondary antibodies that can be used together with minimal cross-reactivity. If the two mouse antibodies are of the same isotype, then you would need to make direct conjugates, such as with our antibody labeling kits or APEX, and SiteClick kits.