Sulfonazo III salt
Sulfonazo III salt
Sulfonazo III salt
Thermo Scientific Chemicals

Sulfonazo III salt

CAS: 68504-35-8 | C22H16N4Na4O14S4 | 780.583 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 A10486.14
亦稱為 A10486-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS68504-35-8
IUPAC Nametetrasodium (3Z,6Z)-4,5-dioxo-3,6-bis[2-(2-sulfonatophenyl)hydrazin-1-ylidene]-3,4,5,6-tetrahydronaphthalene-2,7-disulfonate
Molecular FormulaC22H12N4Na4O14S4
InChI KeyOWVXGCUJHKOBMQ-DTSPTRIMSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1NN=C1C(=O)C2=C(C=C1S([O-])(=O)=O)C=C(C(=N/NC1=CC=CC=C1S([O-])(=O)=O)C2=O)S([O-])(=O)=O
檢視更多
規格Specification Sheet規格表
Formpowder
Loss on Drying< 15%
lambda-max (UV-Vis)2: 562 - 572nm (water)
Specific Absorbance (E 1%/1cm)> 80 @ LMAX1 (water) calculated on dry weight basis
CommentSuitable for SO4 titration: colour change blue to pink
檢視更多
Sulfonazo III salt is used as spectrophotometric reagent for alkaline earth metals and indicator for sulfate titrations. In general it is used as an indicator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sulfonazo III salt is used as spectrophotometric reagent for alkaline earth metals and indicator for sulfate titrations. In general it is used as an indicator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in a cool, dry, well-ventilated area.
RUO – Research Use Only

General References:

  1. Paul J. Kemp; Max B. Williams. Simultaneous spectrophotometric determination of barium and strontium using Sulfonazo III. Anal. Chem.1973, 45, (1) 124-129.
  2. Indicator for determination of sulfates and sulfur: Anal. Chem., 37, 1159 (1965). Spectrophotometric reagent for alkaline earth metals.