Thermo Scientific Chemicals

Chloranilic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10493.22
100 g, Each
Thermo Scientific Chemicals

Chloranilic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A10493.22
100 g, Each
Quantity
Catalog number: A10493.22
also known as A10493-22
Price (USD)
Price: 124.00
Online price: 106.65
Your price:
Quantity
-

Chemical Identifiers

CAS
87-88-7
IUPAC Name
2,5-dichloro-3,6-dihydroxycyclohexa-2,5-diene-1,4-dione
Molecular Formula
C6H2Cl2O4
InChI Key
IPPWILKGXFOXHO-UHFFFAOYSA-N
SMILES
OC1=C(Cl)C(=O)C(O)=C(Cl)C1=O
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Assay (Silylated GC)
≥98.0% (non-U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Water Content (Karl Fischer Titration)
≤2.0%
Appearance (Color)
Red to orange or reddish-orange

Description

Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloranilic acid is used in spectrophotometry. It reacts with metal ions to form stable complexes. It is also used as a reactant in the preparation of dimethylbipyridyl complexes and (nonylbenzimidazolylmethyl)benzene. Further, it is used in charge-transfer reactions with metoprolol tartrate. It is also employed in the synthesis of osmium metalla-rectangles with anticancer activity. In addition to this, it has potential antibacterial activities against methicillin-resistant Staphylococcus aureus.

Solubility
Soluble in hot water and methanol. Insoluble in organic solvents.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text