4-(Trifluoromethyl)benzyl alcohol, 98%
4-(Trifluoromethyl)benzyl alcohol, 98%
4-(Trifluoromethyl)benzyl alcohol, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzyl alcohol, 98%

CAS: 349-95-1 | C8H7F3O | 176.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10508.06
also known as A10508-06
Price (USD)
25.90
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
25.90
Each
Chemical Identifiers
CAS349-95-1
IUPAC Name[4-(trifluoromethyl)phenyl]methanol
Molecular FormulaC8H7F3O
InChI KeyMOOUWXDQAUXZRG-UHFFFAOYSA-N
SMILESOCC1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Trifluoromethyl)benzyl alcohol, is used as a pharmaceutical intermediate, it is also used to predict the NMR spectrum. 4-(Trifluoromethyl)benzyl alcohol is employed as a reagent in, for example, kinetic studies of phosphonoformate prodrugs and aquachromium(IV).

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Keep away from oxidizing agents,reducing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only
T Weber.; J Brunner. 2-(Tributylstannyl)-4-[3-(trifluoromethyl)-3H-diazirin-3-yl] benzyl alcohol: a building block for photolabeling and crosslinking reagents of very high specific radioactivity. J. Am. Chem. Soc. 1995117,(11) , 3084-3095.