Gibco™

Keratinocyte AOF Growth Kit

Catalog number:  A1051501

 Related applications:

Mammalian Cell Culture

| Primary Cell Culture

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Growth kit for the animal origin-free culture of primary human keratinocytes. Contains 1 each of EpiLife Basal Medium (M-EPI-500-CA), Supplement S7 (S-017-5), and Coating Matrix Kit (R-011-K).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type: Keratinocytes
Classification: Animal Origin-Free, Chemically Defined, Serum-Free
Culture Environment: CO2
Culture Type: Adherent Cell Culture
Product Line: Cascade Biologics®
Product Size: 1 kit
Serum Level: Serum-Free
Species: Human

Contents & storage

Contains 1 each of EpiLife Basal Medium (M-EPI-500-CA), Supplement S7 (S-017-5), and Coating Matrix Kit (R-011-K). Store each component at the temperature indicated on the label.
Store each component at the temperature indicated on the label.

Documents

Manuals & protocols