Thermo Scientific Chemicals

Pentaerythritol, 98+%, Thermo Scientific Chemicals

Catalog number: A10520.0I
5000 g, Each
Thermo Scientific Chemicals

Pentaerythritol, 98+%, Thermo Scientific Chemicals

Catalog number: A10520.0I
5000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A10520.0I
also known as A10520-0I
Price (USD)
Price: 124.00
Online price: 105.65
Your price:
Quantity
-

Chemical Identifiers

CAS
115-77-5
IUPAC Name
2,2-bis(hydroxymethyl)propane-1,3-diol
Molecular Formula
C5H12O4
InChI Key
WXZMFSXDPGVJKK-UHFFFAOYSA-N
SMILES
OCC(CO)(CO)CO
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Assay (Silylated GC)
≥98.0%
Water Content (Karl Fischer Titration)
≤1.0%

Description

Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentaerythritol is used as chemical intermediate for alkylated resins, synth lubricants, pentaerythritol tetranitrate & resin & tall oil esters. It is also used in coating, dynamite and plastic industry.

Solubility
Soluble in Water: 0.1 g/mL

Notes
Hygroscopic. Incompatible with strong acids, strong oxidizing agents, acid chlorides, acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text