2,3-Dichlorobenzoic acid, 98%
2,3-Dichlorobenzoic acid, 98%
2,3-Dichlorobenzoic acid, 98%
2,3-Dichlorobenzoic acid, 98%
Thermo Scientific Chemicals

2,3-Dichlorobenzoic acid, 98%

CAS: 50-45-3 | C7H4Cl2O2 | 191.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A10522.18
also known as A10522-18
Price (TWD)
-
Request A Quote
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS50-45-3
IUPAC Name2,3-dichlorobenzoic acid
Molecular FormulaC7H4Cl2O2
InChI KeyQAOJBHRZQQDFHA-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(Cl)=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder and/or Lumps
Assay (Silylated GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only