See more signal from immunofluorescence images with Prolong® Diamond. Learn more.
Invitrogen™

Fluorescein Rabbit Anti-Goat (H+L)

Catalog number:  A10529

Related applications:

Secondary Detection

Back to top

Description

Affinity purified whole antibody rabbit anti-goat IgG conjugated to fluorescein. To minimize cross-reactivity, this antibody has been absorbed against human and mouse serum proteins.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Antibody Conjugate: Classic Dyes (e.g. FITC, TRITC), FITC (Fluorescein)
Antibody Fragment: Whole Antibody
Color: Green
Emission: Visible
Excitation: Visible
Excitation/Emission: 498⁄517
Form: Liquid
Label Type: Classic Dyes, Classic Dyes (e.g. FITC, TRITC)
Label or Dye: FITC (Fluorescein)
Mass: 1 mg
Product Line: Molecular Probes®
Reactivity: Goat
Shipping Condition: Room Temperature
Target Isotype: IgG
Volume (Metric): 0.5 mL
Concentration: 2 mg/mL, 2 mg⁄ml, 2 mg/ml
Host Species: Rabbit

Contents & storage

fluorescein rabbit anti-goat (H+L), 2 mg⁄ml, store at 4°C