What storage conditions should I use for Lithium chloride?
Lithium chloride is hygroscopic and should be stored under a dry, inert atmosphere, and protected from sunlight. Refer to the product label for specific storage temperature requirements.
Search
View additional product information for Lithium chloride, anhydrous, 98+% - FAQs (A10531.22, A10531.36, A10531.0E)
1 product FAQs found