Methyl 3-methyl-2-furoate, 98%
Methyl 3-methyl-2-furoate, 98%
Methyl 3-methyl-2-furoate, 98%
Thermo Scientific Chemicals

Methyl 3-methyl-2-furoate, 98%

CAS: 6141-57-7 | C7H8O3 | 140.14 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
10 g
25 g
產品號碼 A10541.06
亦稱為 A10541-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS6141-57-7
IUPAC Namemethyl 3-methylfuran-2-carboxylate
Molecular FormulaC7H8O3
InChI KeyAQQYRDKMXXSIMP-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(C)C=CO1
檢視更多
規格Specification Sheet規格表
Appearance (Color)White to pale yellow
Assay (GC)≥97.5%
Melting Point (clear melt)31.0-37.0?C
FormCrystals or powder or crystalline powder or fused/lumpy solid
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-methyl-2-furoate is an important raw material used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.It is a volatile substance in the unfermented apple pomace.

Solubility
Insoluble in water.

Notes
Store in a cool dry place. Incompatible with oxidizing agents.
RUO – Research Use Only
George Buechi; Erwin Kovats; P.Enggist; G.Uhde. Synthesis of rosefuran and dehydroelsholtzione. J. Org. Chem. 1968,33, (3),1227-1229