Thermo Scientific Chemicals

Sodium bromide, 99+% (dry wt.), water <1.0%, Thermo Scientific Chemicals

Catalog number: A10552.30
250 g, Each
Thermo Scientific Chemicals

Sodium bromide, 99+% (dry wt.), water <1.0%, Thermo Scientific Chemicals

Catalog number: A10552.30
250 g, Each
Quantity
Catalog number: A10552.30
also known as A10552-30
Price (USD)
Price: 37.30
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7647-15-6
IUPAC Name
sodium bromide
Molecular Formula
BrNa
InChI Key
JHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES
[Na+].[Br-]
Form
Crystals or powder or crystalline powder
Appearance (Color)
White
Assay (Titration ex Bromide)
≥99.0 to ≤101.0% (dry wt. basis)
Water Content (Karl Fischer Titration)
≤1%

Description

Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium bromide is used in pharmaceutical applications, used in photographic processing and chemical intermediate for production of chemicals and bromides. It is widely used as an anticonvulsant. Also used for the preparation of drilling fluids, completion fluids and drilling muds formulations.

Solubility
Soluble in water, soluble in alcohol.

Notes
It is thermally and chemically stable. Hygroscopic. Sodium bromide is non-flammable and when strongly heated it may emit bromine fumes.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text