Iodomethane, 99%, stab. with copper
Iodomethane, 99%, stab. with copper
Iodomethane, 99%, stab. with copper
Thermo Scientific Chemicals

Iodomethane, 99%, stab. with copper

CAS: 74-88-4 | CH3I | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number A10559.30
also known as A10559-30
Price (EUR)
128,65
Online Exclusive
143,00
Save 14,35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
128,65
Online Exclusive
143,00
Save 14,35 (10%)
Each
Chemical Identifiers
CAS74-88-4
IUPAC Nameiodomethane
Molecular FormulaCH3I
InChI KeyINQOMBQAUSQDDS-UHFFFAOYSA-N
SMILESCI
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥98.5% (U.S. specification)
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.5280-1.5330 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to yellow to orange or pale pink
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodomethane is a methylating agent used for alkylating carbon, oxygen, sulfur, nitrogen and phosphorus nucleophiles. For example, phenol reacts with methyl iodide to give anisole. In the Monsanto process, the insitu formed iodomethane reacts with carbon monoxide in the presence of a rhodium complex to give acetyl iodide, which on hydrolysis yields acetic acid. It reacts with magnesium to form the Grignard reagent, methylmagnesium iodide used in organic synthesis. It finds applications as an intermediate in the manufacture of pharmaceuticals and in phase transfer catalysts.

Solubility
Miscible with water.

Notes
Light and moisture sensitive. Store in cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and oxygen.
RUO – Research Use Only

General References:

  1. Erk, B.; Boll, R., Trippel, S.; Anielski, D.; Foucar, L.; Rudek, B.; Rudenko, A. Imaging charge transfer in iodomethane upon x-ray photoabsorption. Science 2014, 345 (6194), 288-291.
  2. Dong, Y.; Brooks, J. D.; Chen, T. L.; Mullins, D. R.; Cox, D. F. Reactions of methyl groups on a non-reducible metal oxide: The reaction of iodomethane on stoichiometric alpha-Cr 2 O 3 (0001). Surf. Sci. 2015, 641, 148-153.