Ácido 4-hidroxi-3-nitrobenzoico, 98 %, Thermo Scientific Chemicals
Ácido 4-hidroxi-3-nitrobenzoico, 98 %, Thermo Scientific Chemicals
Ácido 4-hidroxi-3-nitrobenzoico, 98 %, Thermo Scientific Chemicals
Ácido 4-hidroxi-3-nitrobenzoico, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido 4-hidroxi-3-nitrobenzoico, 98 %, Thermo Scientific Chemicals

CAS: 616-82-0 | C7H5NO5 | 183.12 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Número de catálogo A10562.22
también denominado A10562-22
Precio (USD)
-
Cantidad:
100 g
Pedido a granel o personalizado
Identificadores químicos
CAS616-82-0
IUPAC Name4-hydroxy-3-nitrobenzoic acid
Molecular FormulaC7H5NO5
InChI KeyQRYSWXFQLFLJTC-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(O)C(=C1)[N+]([O-])=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to cream to pale yellow
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)179.0-185.0?C
FormPowder
Assay (Silylated GC)≥97.5%
Standard in chymotrypsin assays

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only