2,6-Dibromophenol, 99%
2,6-Dibromophenol, 99%
2,6-Dibromophenol, 99%
Thermo Scientific Chemicals

2,6-Dibromophenol, 99%

CAS: 608-33-3 | C6H4Br2O | 251.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A10573.14
also known as A10573-14
Price (EUR)
431,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
431,00
Each
Chemical Identifiers
CAS608-33-3
IUPAC Name2,6-dibromophenol
Molecular FormulaC6H4Br2O
InChI KeySSIZLKDLDKIHEV-UHFFFAOYSA-N
SMILESOC1=C(Br)C=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale grey
Melting Point (clear melt)51.0-58.0?C
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
2,6-Dibromophenol is used in the production of 3,4,5-trimethoxybenzaldehyde and 2-(2,6-dibromophenoxy)ethylbromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromophenol is used in the production of 3,4,5-trimethoxybenzaldehyde and 2-(2,6-dibromophenoxy)ethylbromide.

Solubility
Soluble in ethanol and ether. Slightly soluble in water.

Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents.
RUO – Research Use Only

General References:

  1. Dolfing, J.; Novak, I. The Gibbs free energy of formation of halogenated benzenes, benzoates and phenols and their potential role as electron acceptors in anaerobic environments. Biodegradation 2015, 26 (1), 15-27.
  2. Chen, K.; Jian, S.; Huang, L.; Ruan, Z.; Li, S.; Jiang, J. Reductive dehalogenation of 3,5-dibromo-4-hydroxybenzoate by an aerobic strain of Delftia sp. EOB-17. Biotechnol. Lett 2015, 37 (12), 2395-2401.