Thermo Scientific Chemicals

Methyl salicylate, 98%, Thermo Scientific Chemicals

Catalog number: A10578.36
500 g, Each
Thermo Scientific Chemicals

Methyl salicylate, 98%, Thermo Scientific Chemicals

Catalog number: A10578.36
500 g, Each
Quantity
Catalog number: A10578.36
also known as A10578-36
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
119-36-8
IUPAC Name
methyl 2-hydroxybenzoate
Molecular Formula
C8H8O3
InChI Key
OSWPMRLSEDHDFF-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=CC=C1O
Refractive Index
1.5335-1.5375 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Identification (FTIR)
Conforms
Assay (GC)
≥97.5%
Form
Liquid

Description

Methyl salicylate is used as a flavoring agent and providing fragrance to products like chewing gum and mints. It acts as an anticoagulant by interruption of the vitamin K1-epoxide cycle. It is also used in high concentrations as a rubefacient to treat joint, muscular pain and acute pain. It is utilized as an anti-herbivore defense system in various plants. It is used as a transfer agent to get a manual copy of an image on a surface and to clear plant or animal tissue samples to be used for microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl salicylate is used as a flavoring agent and providing fragrance to products like chewing gum and mints. It acts as an anticoagulant by interruption of the vitamin K1-epoxide cycle. It is also used in high concentrations as a rubefacient to treat joint, muscular pain and acute pain. It is utilized as an anti-herbivore defense system in various plants. It is used as a transfer agent to get a manual copy of an image on a surface and to clear plant or animal tissue samples to be used for microscopy.

Solubility
Miscible with water, chloroform, ethanol, methonol and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Acute oral toxicity
    Category 4
    Serious Eye Damage/Eye Irritation
    Category 1
    Skin Sensitization
    Category 1
    Skin Sensitization
    Sub-category 1B
    Reproductive Toxicity
    Category 2
    Chronic aquatic toxicity
    Category 3
    Label Elements
    Signal Word

    Danger

    Hazard Statements

    H302 - Harmful if swallowed

    H317 - May cause an allergic skin reaction

    H318 - Causes serious eye damage

    H361d - Suspected of damaging the unborn child

    H412 - Harmful to aquatic life with long lasting effects

    Precautionary Statements

    P280 - Wear protective gloves/protective clothing/eye protection/face protection

    P301 + P330 + P331 - IF SWALLOWED: rinse mouth. Do NOT induce vomiting

    P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

    P305 + P351 + P338 - IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing

    P310 - Immediately call a POISON CENTER or doctor/physician