Thermo Scientific Chemicals

Nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A10585.0E
2500 g, Each
Thermo Scientific Chemicals

Nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog number: A10585.0E
2500 g, Each
Quantity
Catalog number: A10585.0E
also known as A10585-0E
Price (USD)
Price: 114.00
Your price:
Quantity
-

Chemical Identifiers

CAS
98-95-3
IUPAC Name
nitrobenzene
Molecular Formula
C6H5NO2
InChI Key
LQNUZADURLCDLV-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC=C1
Appearance (Color)
Clear colorless to yellow
Form
Liquid
Assay (GC)
≥98.5%
Identification (FTIR)
Conforms
Water Content (Karl Fischer Titration)
≤0.15%

Description

Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for electrophilic reagents in synthetic chemistry. Further, it is used to prevent the unpleasant odor in shoe, floor and metal polishes. It acts as a precursor to rubber chemicals, azo dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nitrobenzene is used as a solvent and as a starting material in the organic synthesis for the preparation of aniline, benzidine, azobenzene, nitrosobenzene, phenylhydroxylamine and acetaminophen. It is also used as a solvent for electrophilic reagents in synthetic chemistry. Further, it is used to prevent the unpleasant odor in shoe, floor and metal polishes. It acts as a precursor to rubber chemicals, azo dyes and pharmaceuticals.

Solubility
Miscible with ethanol, diethyl ether, acetone, benzene and organic solvents. Slightly miscible with water and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text