2-苯基苯酚, 99%
2-苯基苯酚, 99%
2-苯基苯酚, 99%
Thermo Scientific Chemicals

2-苯基苯酚, 99%

CAS: 90-43-7 | C12H10O | 170.21 g/mol
Have Questions?
更改视图buttonViewtableView
数量:
250 g
500 g
2500 g
货号 A10592.0E
又称 A10592-0E
价格(TWD)
-
数量:
2500 g
请求批量或定制报价
化学标识符
CAS90-43-7
IUPAC Name[1,1'-biphenyl]-2-ol
Molecular FormulaC12H10O
InChI KeyLLEMOWNGBBNAJR-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C1=CC=CC=C1
查看更多内容
规格Specification Sheet规格表
Appearance (Color)White to pale pink
Assay (GC)≥98.5%
Melting Point (clear melt)54.0-60.0?C
FormCrystals or powder or crystalline powder or lumps or flakes
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Phenylphenol is used as a dye intermediate, plasticizer, rubber chemical and as a preservative in water-oil emulsions. It is used as an antimicrobial preservative in cosmetics and an antimicrobial additive in the manufacture of metalworking fluids, leather, adhesives and textiles. Further, it serves as a disinfectant in households, hospitals, nursing homes, farms, laundries, barber shops, veterinary equipment and food processing plants. It is also employed in the production of resins and rubber chemicals. In addition to this, it is used as a polymer material, stabilizer and flame retardant.

Solubility
Soluble in ethanol, acetone, benzene,sodium hydroxide, chloroform, acetonitrile, toluene, hexane, ligroin, ethyl ether, pyridine, ethylene glycol, isopropanol, glycol ethers and polyglycols.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Seki, Y.; Ide, Y.; Okada, T.; Ogawa, M. Concentration of 2-phenylphenol by organoclays from aqueous sucrose solution. Appl. Clay Sci. 2015, 109-110, 64-67.
  2. Ma, J.; Zhang, X.; Basarić, N.; Wan, P.; Phillips, D. L. Observation of excited state proton transfer reactions in 2-phenylphenol and 2-phenyl-1-naphthol and formation of quinone methide species. Phys. Chem. Chem. Phys. 2015, 17 (14), 9205-9211.