Thermo Scientific Chemicals

Anisil, 98+%, Thermo Scientific Chemicals

Catalog number: A10594.22
100 g, Each
Thermo Scientific Chemicals

Anisil, 98+%, Thermo Scientific Chemicals

Catalog number: A10594.22
100 g, Each
Quantity
Catalog number: A10594.22
also known as A10594-22
Price (USD)
Price: 740.00
Online price: 639.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1226-42-2
IUPAC Name
bis(4-methoxyphenyl)ethane-1,2-dione
Molecular Formula
C16H14O4
InChI Key
YNANGXWUZWWFKX-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)C(=O)C(=O)C1=CC=C(OC)C=C1
Melting Point (clear melt)
131-135?C
Appearance (Color)
Yellow
Assay (GC)
≥98.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms

Description

Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anisil is a biochemical utilized in the preparation of substituted bis(dithiolene)nickel complex. Further, it is used in the preparation of 3-[4,5-bis-(4-methoxy-phenyl)-1H-imidazol-2-yl]-pyridine by reacting with pyridine-3-carbaldehyde.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text