2,3,4-Trimethoxybenzaldehyde, 98+%
2,3,4-Trimethoxybenzaldehyde, 98+%
2,3,4-Trimethoxybenzaldehyde, 98+%
Thermo Scientific Chemicals

2,3,4-Trimethoxybenzaldehyde, 98+%

CAS: 2103-57-3 | C10H12O4 | 196.202 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number A10609.18
also known as A10609-18
Price (USD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS2103-57-3
IUPAC Name2,3,4-trimethoxybenzaldehyde
Molecular FormulaC10H12O4
InChI KeyUCTUXUGXIFRVGX-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(OC)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (GC)≥98.0%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or fused solid
Free acid (titration)≤1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only