1,3-Diiodopropane, 98+%, stab. with copper
1,3-Diiodopropane, 98+%, stab. with copper
1,3-Diiodopropane, 98+%, stab. with copper
Thermo Scientific Chemicals

1,3-Diiodopropane, 98+%, stab. with copper

CAS: 627-31-6 | C3H6I2 | 295.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number A10616.30
also known as A10616-30
Price (EUR)
345,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
345,00
Each
Chemical Identifiers
CAS627-31-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (GC)≥98.0%
Refractive Index1.6395-1.6435 @ 20?C
FormLiquid
Identification (FTIR)Conforms
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Stable, but light sensitive - store in the dark. Incompatible with strong bases, strong oxidizing agents.
RUO – Research Use Only

General References:

  1. J Thorbjørnsrud.; OH Ellestad.; P Klaboe. Substituted propanes: VI. The vibrational spectra and conformations of 1, 3-dichloro-, 1, 3-bromochloro-, 1, 3-dibromo-and 1, 3-diiodo-propane. Journal of Molecular Structure. 1973 15, (1) , 61-76.
  2. GA Crowder.; S Ali. Vibrational analysis of 1-iodopropane, 1-iodobutane and 1, 3-diiodopropane. Journal of Molecular Structure. 1975 25, (2) , 377-386.