5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
RUO – Research Use Only
General References:
- ER Pérez.; AL Marrero.; R Pérez.; MA Autié. An efficient microwave-assisted method to obtain 5-nitrofurfural without solvents on mineral solid supports. Tetrahedron letters. 1995 36, (11) ,1779-1782.
- RW Busker.; GMJB van Henegouwen.; RF Menke. Formation of methemoglobin by photoactivation of nitrofurantoin or of 5-nitrofurfural in rats exposed to UV-A light. Toxicology. 1995 51, (2-3) , 255-266.