2-Chloroveratraldehyde, 98%, Thermo Scientific Chemicals
2-Chloroveratraldehyde, 98%, Thermo Scientific Chemicals
2-Chloroveratraldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroveratraldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
A10623.14
also known as A10623-14
25 g
Catalog number A10623.14
also known as A10623-14
Price (EUR)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS5417-17-4
IUPAC Name2-chloro-3,4-dimethoxybenzaldehyde
Molecular FormulaC9H9ClO3
InChI KeySAWHDJTZESXNMM-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(Cl)=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to yellow to brown
Assay (GC)≥97.5%
Free acid (titration)≤1%
FormPowder
Melting Point (clear melt)68.0-74.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only