Thermo Scientific Chemicals

3-Nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10629.22
100 g, Each
Thermo Scientific Chemicals

3-Nitroaniline, 98%, Thermo Scientific Chemicals

Catalog number: A10629.22
100 g, Each
Quantity
Catalog number: A10629.22
Price (USD)
Price: 38.60
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
99-09-2
IUPAC Name
3-nitroaniline
Molecular Formula
C6H6N2O2
InChI Key
XJCVRTZCHMZPBD-UHFFFAOYSA-N
SMILES
NC1=CC=CC(=C1)[N+]([O-])=O
Form
Powder
Melting Point (clear melt)
110.0-116.0?C
Appearance (Color)
Yellow to orange
Assay (GC)
≥97.5%

Description

3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitroaniline is commonly used as a raw material for dyes. It is also used as a chemical intermediate for azo coupling component 17 and the dyes disperse yellow 5 and acid blue 29. The chemical is changed to other substances (dyestuffs and m-nitrophenol) during the dyeing process.

Solubility
It is soluble in water 0.1 g/100 ml (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from acids, acid chlorides, acid anhydrides, chloroformates, strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text