alpha-Benzoin oxime, 98+%
alpha-Benzoin oxime, 98+%
alpha-Benzoin oxime, 98+%
Thermo Scientific Chemicals

alpha-Benzoin oxime, 98+%

CAS: 441-38-3 | C14H13NO2 | 227.26 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
產品號碼 A10630.36
亦稱為 A10630-36
價格 (TWD)
-
Quantity:
500 g
Request bulk or custom format
化學識別
CAS441-38-3
IUPAC Name(2E)-2-(N-hydroxyimino)-1,2-diphenylethan-1-ol
Molecular FormulaC14H13NO2
InChI KeyWAKHLWOJMHVUJC-FYWRMAATNA-N
SMILESO\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Assay (Silylated GC)≥98.0%
Appearance (Color)White to pale cream to pale brown
Melting Point (clear melt)147.0-154.0?C
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only