MembraneMax™ Protein Expression Kit - FAQs
MembraneMax™ Protein Expression Kit - FAQs
View additional product information for MembraneMax™ Protein Expression Kit - FAQs (A10632)
1 product FAQs found
What are the advantages of using a cell-free expression system?
A cell-free expression system is best used when working with a toxic target, as no cells are needed for protein expression. In vitro protein expression utilizes the necessary cellular components to drive expression in a single tube.
Find additional tips, troubleshooting help, and resources within our Protein Expression Support Center.