3-Thiopheneacetic acid, 98%
3-Thiopheneacetic acid, 98%
3-Thiopheneacetic acid, 98%
Thermo Scientific Chemicals

3-Thiopheneacetic acid, 98%

CAS: 6964-21-2 | C6H6O2S | 142.172 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10700.14
also known as A10700-14
Price (EUR)
189,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
189,00
Each
Chemical Identifiers
CAS6964-21-2
IUPAC Name2-(thiophen-3-yl)acetic acid
Molecular FormulaC6H6O2S
InChI KeyRCNOGGGBSSVMAS-UHFFFAOYSA-N
SMILESOC(=O)CC1=CSC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)77.0-83.0?C
Appearance (Color)Pale cream to pale brown
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
View more
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Thiopheneacetic acid was used in one-step, size control synthesis of gold nanoparticles. It was also used in the preparation of gold nanoparticles capped with 3-thiopheneacetic acid (3-TAA) via borohydride reduction.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

General References:

  1. Csaba Janaky; Csaba Visy; Otto Berkesi; Etelka Tombacz. Conducting Polymer-Based Electrode with Magnetic Behavior: Electrochemical Synthesis of Poly(3-thiophene-acetic-acid)/Magnetite Nanocomposite Thin Layer. J. Phys. Chem. C 2009, 113, (4),1352-1358
  2. M.Aydin; Z.Durmus; H.Kavas; B.Esat; H.Sozeri; A.Baykal; F.Yilmaz; M.S.Toprak. Synthesis and characterization of poly(3-thiophene acetic acid)/Fe3O4 nanocomposite. Polyhedron. 2011,30, (6),1120-1126