3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ca 10% water, Thermo Scientific Chemicals
3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ca 10% water, Thermo Scientific Chemicals
3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ca 10% water, Thermo Scientific Chemicals
3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ca 10% water, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,5-Diiodo-L-tyrosine hydrate, 98% (dry wt.), may contain up to ca 10% water, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A10702.22
also known as A10702-22
Price (TWD)
-
Request A Quote
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS18835-59-1
IUPAC Name(2S)-2-amino-3-(4-hydroxy-3,5-diiodophenyl)propanoic acid dihydrate
Molecular FormulaC9H13I2NO5
InChI KeyYWAGQOOMOOUEGY-QVGJJETENA-N
SMILESO.O.N[C@@H](CC1=CC(I)=C(O)C(I)=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (Aqueous acid-base Titration)≥97.5% (non-U.S. sourced material)
Assay from Supplier's CofA≥97.5% (U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only